Assay Details
Target Gene Details
Entrez Gene ID: | 112399 |
Gene Name: | egl-9 family hypoxia inducible factor 3 |
Gene Aliases: |
HIFP4H3, HIFPH3, PHD3 |
Location: |
Chr.14:33924215-33951078 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGLN3 | NM_001308103.1 | NP_001295032.1 | ||
NM_022073.3 | NP_071356.1 | |||
AJ310545.1 | CAC42511.1 | |||
AK025273.1 | BAB15101.1 | |||
AK123350.1 | ||||
AK225473.1 | ||||
BC010992.2 | ||||
BC064924.1 | ||||
BC102030.1 | AAI02031.1 | |||
BC105938.1 | AAI05939.1 | |||
BC105939.1 | AAI05940.1 | |||
BC111057.1 | ||||
BG716229.1 | ||||
DQ975379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1070213 | Chr.14:33930300 - 33936238 on Build GRCh38 | Deletion | EGLN3 |
nsv1036996 | Chr.14:33834296 - 34189902 on Build GRCh38 | Gain | EGLN3-AS1 EGLN3 |
esv34070 | Chr.14:33763566 - 34160397 on Build GRCh38 | Loss | EGLN3-AS1 EGLN3 NPAS3 |
nsv832768 | Chr.14:33887269 - 34082696 on Build GRCh38 | Gain | EGLN3-AS1 EGLN3 |
nsv832769 | Chr.14:33927540 - 34114539 on Build GRCh38 | Loss | EGLN3-AS1 EGLN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |