Assay Details
Target Gene Details
Entrez Gene ID: | 883 |
Gene Name: | kynurenine aminotransferase 1 |
Gene Aliases: |
CCBL1, GTK, KAT1, KATI |
Location: |
Chr.9:128832942-128882551 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KYAT1 | NM_001122671.1 | NP_001116143.1 | ||
NM_001122672.1 | NP_001116144.1 | |||
NM_001287390.1 | NP_001274319.1 | |||
NM_004059.4 | NP_004050.3 | |||
NR_109829.1 | ||||
XM_006717311.3 | XP_006717374.1 | |||
XM_011519165.2 | XP_011517467.1 | |||
XM_011519167.2 | XP_011517469.2 | |||
XM_011519169.1 | XP_011517471.1 | |||
XM_017015260.1 | XP_016870749.1 | |||
XM_017015261.1 | XP_016870750.1 | |||
XM_017015263.1 | XP_016870752.1 | |||
XM_017015264.1 | XP_016870753.1 | |||
XM_017015266.1 | XP_016870755.1 | |||
XM_017015267.1 | XP_016870756.1 | |||
XM_017015268.1 | XP_016870757.1 | |||
AK291178.1 | ||||
AK297995.1 | ||||
AK314427.1 | ||||
AU123740.1 | ||||
BC033685.1 | AAH33685.1 | |||
BI755163.1 | ||||
DA381449.1 | ||||
DC413592.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |