Assay Details
Target Gene Details
Entrez Gene ID: | 121278 |
Gene Name: | tryptophan hydroxylase 2 |
Gene Aliases: |
ADHD7, NTPH |
Location: |
Chr.12:71938846-72032441 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPH2 | NM_173353.3 | NP_775489.2 | ||
AK094614.1 | BAC04385.1 | |||
AY098914.1 | AAM28946.1 | |||
BC114442.1 | ||||
BC114499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518498 | Chr.12:71913836 - 72114999 on Build GRCh38 | Loss | TBC1D15 TRHDE TPH2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |