Assay Details
Target Gene Details
Entrez Gene ID: | 114899 |
Gene Name: | C1q and tumor necrosis factor related protein 3 |
Gene Aliases: |
C1ATNF3, CORCS, CORS, CORS-26, CORS26, CTRP3 |
Location: |
Chr.5:34017858-34043266 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1QTNF3 | NM_030945.3 | NP_112207.1 | ||
NM_181435.5 | NP_852100.3 | |||
AF173888.1 | AAQ13635.1 | |||
AF326976.1 | AAK70344.1 | |||
AF329837.1 | AAK17961.1 | |||
AK075533.1 | BAC11676.1 | |||
AK295968.1 | ||||
AK315921.1 | ||||
AY358388.1 | AAQ88754.1 | |||
BC112925.1 | ||||
BC120990.2 | ||||
BX640995.1 | CAE45998.1 | |||
EU399231.1 | ||||
EU399232.1 |
Target Gene Details
Entrez Gene ID: | 100534612 |
Gene Name: | C1QTNF3-AMACR readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.5:33986986-34124528 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1QTNF3-AMACR | NR_037951.1 | |||
AB209261.1 | BAD92498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830255 | Chr.5:33929507 - 34109160 on Build GRCh38 | Loss | C1QTNF3-AMACR RXFP3 SLC45A2 AMACR C1QTNF3 |
nsv519240 | Chr.5:33950598 - 34749311 on Build GRCh38 | Gain | LOC105374753 C1QTNF3-AMACR LOC646652 RAI14 SLC45A2 AMACR C1QTNF3 |
nsv522715 | Chr.5:33319157 - 34037875 on Build GRCh38 | Gain | TARS ADAMTS12 C1QTNF3-AMACR RXFP3 SLC45A2 AMACR C1QTNF3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |