Assay Details
Target Gene Details
Entrez Gene ID: | 239 |
Gene Name: | arachidonate 12-lipoxygenase, 12S type |
Gene Aliases: |
12-LOX, 12S-LOX, LOG12 |
Location: |
Chr.17:6993791-7010736 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALOX12 | NM_000697.2 | NP_000688.2 | ||
XM_011523780.2 | XP_011522082.1 | |||
BC069557.1 | AAH69557.1 | |||
M35418.1 | AAA60056.1 | |||
M58704.1 | AAA59523.1 | |||
M62982.1 | AAA51533.1 | |||
S68587.1 | AAD14020.1 | |||
S68588.1 | AAD14021.1 |
Target Gene Details
Entrez Gene ID: | 100506713 |
Gene Name: | ALOX12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:6985123-7012334 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALOX12-AS1 | NR_040089.1 | |||
BI559919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | TEKT1 GABARAP PLSCR3 MIR195 ACADVL SNORA67 ATP1B2 FBXO39 MPDU1 EIF4A1 CHRNB1 SAT2 SENP3-EIF4A1 SLC16A13 ALOX12P2 SLC16A11 ZBTB4 MIR497HG CLDN7 TNFSF13 POLR2A TMEM102 TNK1 TNFSF12 CD68 KCTD11 PHF23 SLC35G6 NLGN2 CTDNEP1 LOC107983988 RNASEK-C17orf49 SNORA48 TMEM256 MIR497 CLEC10A DLG4 DVL2 SOX15 YBX2 SENP3 GPS2 RNASEK C17orf49 LOC100996842 ACAP1 FGF11 TMEM95 ALOX12 TMEM256-PLSCR3 SPEM1 ALOX12-AS1 C17orf74 ASGR1 SNORD10 MIR324 SLC2A4 EIF5A ASGR2 BCL6B ELP5 FXR2 NEURL4 SHBG TNFSF12-TNFSF13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |