Assay Details
Target Gene Details
Entrez Gene ID: | 112817 |
Gene Name: | 4-hydroxy-2-oxoglutarate aldolase 1 |
Gene Aliases: |
C10orf65, DHDPS2, DHDPSL, HP3, NPL2 |
Location: |
Chr.10:97584345-97612802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HOGA1 | NM_001134670.1 | NP_001128142.1 | ||
NM_138413.3 | NP_612422.2 | |||
AK094791.1 | ||||
AK289440.1 | ||||
BC011916.2 | AAH11916.1 | |||
BC045550.1 | AAH45550.1 | |||
BC057821.1 | AAH57821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047359 | Chr.10:97546744 - 97609256 on Build GRCh38 | Gain | C10orf62 HOGA1 UBTD1 ANKRD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |