Assay Details
Target Gene Details
Entrez Gene ID: | 7433 |
Gene Name: | vasoactive intestinal peptide receptor 1 |
Gene Aliases: |
HVR1, II, PACAP-R-2, PACAP-R2, RDC1, V1RG, VAPC1, VIP-R-1, VIPR, VIRG, VPAC1, VPAC1R, VPCAP1R |
Location: |
Chr.3:42489299-42537573 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VIPR1 | NM_001251882.1 | NP_001238811.1 | ||
AK056819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527430 | Chr.3:42492550 - 42515707 on Build GRCh38 | Loss | VIPR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |