Assay Details
Target Gene Details
Entrez Gene ID: | 51661 |
Gene Name: | FK506 binding protein 7 |
Gene Aliases: |
FKBP23, PPIase |
Location: |
Chr.2:178463664-178479878 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP7 | NM_001135212.1 | NP_001128684.1 | ||
NM_181342.2 | NP_851939.1 | |||
XM_005246638.4 | XP_005246695.1 | |||
XM_011511348.2 | XP_011509650.1 | |||
XM_011511349.2 | XP_011509651.1 | |||
AF092137.1 | AAD40379.1 | |||
AF100751.1 | AAD43015.1 | |||
AK292145.1 | ||||
AK299215.1 | ||||
AY353086.1 | AAQ57208.1 | |||
AY359015.1 | AAQ89374.1 | |||
BC009711.1 | AAH09711.1 | |||
BF210667.1 | ||||
BT007122.1 | AAP35786.1 | |||
DA020387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss | DFNB59 PRKRA TTN-AS1 FKBP7 TTN PLEKHA3 LOC101927027 OSBPL6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |