Assay Details
Target Gene Details
Entrez Gene ID: | 51639 |
Gene Name: | splicing factor 3b subunit 6 |
Gene Aliases: |
CGI-110, HSPC175, Ht006, P14, SAP14, SAP14a, SF3B14, SF3B14a |
Location: |
Chr.2:24067584-24076444 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SF3B6 | NM_016047.3 | NP_057131.1 | ||
AF151868.1 | AAD34105.1 | |||
AF161523.1 | AAF29138.1 | |||
AF184213.1 | AAG09698.1 | |||
AF401310.1 | AAK94041.1 | |||
AK000621.1 | ||||
AK311118.1 | ||||
AK311908.1 | ||||
BC015463.1 | AAH15463.1 | |||
BG200650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain | MFSD2B FAM228A SF3B6 TP53I3 PFN4 LOC105374325 UBXN2A WDCP KLHL29 FAM228B FKBP1B ITSN2 ATAD2B |
nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain | PTRHD1 FAM228A EFR3B SF3B6 ADCY3 TP53I3 PFN4 CENPO FAM228B FKBP1B DNAJC27-AS1 DNAJC27 ITSN2 NCOA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |