Assay Details
Target Gene Details
Entrez Gene ID: | 130589 |
Gene Name: | galactose mutarotase |
Gene Aliases: |
BLOCK25, GLAT, HEL-S-63p, IBD1 |
Location: |
Chr.2:38665910-38734767 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALM | NM_138801.2 | NP_620156.1 | ||
XM_011532540.1 | XP_011530842.1 | |||
AK291489.1 | ||||
BC014916.1 | AAH14916.1 | |||
BC019263.1 | AAH19263.1 | |||
DA984992.1 | ||||
EU794611.1 | ||||
U11036.1 | AAA67652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3590419 | Chr.2:38660017 - 38726534 on Build GRCh38 | Loss | GALM |
esv3891860 | Chr.2:38666518 - 38724524 on Build GRCh38 | Loss | GALM |
nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss | DHX57 SRSF7 LOC101929596 HNRNPLL GEMIN6 ATL2 GALM CYP1B1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |