Assay Details
Target Gene Details
Entrez Gene ID: | 656 |
Gene Name: | bone morphogenetic protein 8b |
Gene Aliases: |
BMP8, OP2 |
Location: |
Chr.1:39757182-39788861 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BMP8B | NM_001720.3 | NP_001711.2 | ||
XM_005271149.4 | XP_005271206.1 | |||
XM_011542022.2 | XP_011540324.1 | |||
XM_011542024.2 | XP_011540326.2 | |||
XM_011542025.2 | XP_011540327.1 | |||
XM_017002155.1 | XP_016857644.1 | |||
XM_017002156.1 | XP_016857645.1 | |||
AY303955.1 | AAP74560.1 | |||
BC108678.1 | AAI08679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss | TRIT1 PPIE BMP8B PPT1 RLF MFSD2A OXCT2 LOC101929536 CAP1 MYCL |
dgv152n100 | Chr.1:39693156 - 39800418 on Build GRCh38 | Gain | PPIE BMP8B OXCT2 LOC101929536 |
dgv153n100 | Chr.1:39715819 - 39848248 on Build GRCh38 | Gain | TRIT1 PPIE BMP8B OXCT2 LOC101929536 |
nsv546063 | Chr.1:39719061 - 39841226 on Build GRCh38 | Gain | TRIT1 PPIE BMP8B OXCT2 LOC101929536 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |