Assay Details
Target Gene Details
Entrez Gene ID: | 115 |
Gene Name: | adenylate cyclase 9 |
Gene Aliases: |
AC9, ACIX |
Location: |
Chr.16:3953387-4116185 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY9 | XM_011522353.2 | XP_011520655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv21490 | Chr.16:3825338 - 4270692 on Build GRCh38 | Gain+Loss | LOC102724927 ADCY9 LINC01569 SRL TFAP4 LOC105371063 CREBBP |
nsv833131 | Chr.16:3955868 - 4135556 on Build GRCh38 | Loss | ADCY9 |
nsv1038196 | Chr.16:3917410 - 4153984 on Build GRCh38 | Gain | LOC102724927 ADCY9 |
nsv833130 | Chr.16:3810372 - 3975509 on Build GRCh38 | Gain | LOC102724927 ADCY9 CREBBP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |