Assay Details
Target Gene Details
Entrez Gene ID: | 11051 |
Gene Name: | nudix hydrolase 21 |
Gene Aliases: |
CFIM25, CPSF5 |
Location: |
Chr.16:56429136-56451349 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT21 | NM_007006.2 | NP_008937.1 | ||
AJ001810.1 | CAA05026.1 | |||
AK312721.1 | ||||
AL535307.3 | ||||
BC001403.2 | AAH01403.1 | |||
BX537360.1 | CAD97606.1 | |||
BX647665.1 | CAI46057.1 | |||
CR456919.1 | CAG33200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833242 | Chr.16:56259354 - 56446840 on Build GRCh38 | Loss | GNAO1 NUDT21 LOC102723466 AMFR |
nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain | BBS2 OGFOD1 NUDT21 AMFR |
esv3638695 | Chr.16:56413924 - 56440936 on Build GRCh38 | Gain | NUDT21 AMFR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |