Assay Details
Target Gene Details
Entrez Gene ID: | 4883 |
Gene Name: | natriuretic peptide receptor 3 |
Gene Aliases: |
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC |
Location: |
Chr.5:32689076-32791724 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPR3 | XM_011514047.1 | XP_011512349.1 | ||
XM_011514048.2 | XP_011512350.1 | |||
XM_011514049.2 | XP_011512351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017141 | Chr.5:32615443 - 32722393 on Build GRCh38 | Gain | NPR3 |
esv3604616 | Chr.5:32625459 - 32725468 on Build GRCh38 | Gain | NPR3 |
esv2759338 | Chr.5:32245255 - 32738538 on Build GRCh38 | Loss | ZFR NPR3 MTMR12 SUB1 MIR579 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |