Assay Details
Target Gene Details
Entrez Gene ID: | 23583 |
Gene Name: | single-strand-selective monofunctional uracil-DNA glycosylase 1 |
Gene Aliases: |
FDG, HMUDG, UNG3 |
Location: |
Chr.12:54171815-54188994 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509467 | Chr.12:54173617 - 54255598 on Build GRCh38 | Insertion | CBX5 SMUG1 MIR3198-2 |
nsv832419 | Chr.12:54064043 - 54259156 on Build GRCh38 | Loss | LOC100240735 CBX5 FLJ12825 LOC400043 SMUG1 LOC100240734 MIR3198-2 |
nsv832420 | Chr.12:54137272 - 54296797 on Build GRCh38 | Loss | CBX5 HNRNPA1 NFE2 SMUG1 MIR3198-2 |
nsv832418 | Chr.12:54016556 - 54186660 on Build GRCh38 | Loss | LOC100240735 FLJ12825 LOC400043 SMUG1 MIR615 LOC100240734 HOXC5 HOXC4 HOXC6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |