Fisherbrand Microscopy Solutions
Upright, Stereo, and Inverted Microscopes
Browse these products:
Fisherbrand Entry Level Research Grade Inverted Microscope
Perfect for classroom observations of living cells and organisms under more natural conditions.
- Siedentopf head design for more flexible eyepiece adjustment
- Infinity-corrected optics for consistency between objectives
- Extra-long working distance (up to 195mm)
- Koehler LED illumination for uniform and glare-free viewing
Fisherbrand Research Grade Upright Microscopes are available in a number of models and with a range of features.
Choose advanced models for photography and more demanding research, research models for most clinical and research lab use, or entry-level models for general laboratory demands.
Zoom stereo microscopes are available in research grade or entry level, with corresponding features and options. Or choose an entry-level stereo microscope with dual, fixed magnification for more routine use.
Cameras and Camera Mounts
Capture images with trinocular model microscopes, add a camera mount, and find a camera with the right resolution to support your research.
Microscope Eyepieces
Choose from a wide range of eyepieces to accommodate various users and magnifications demands.
Microscope Objectives
Add new or replace your microscope objectives as your research needs and applications change.
Fisherbrand Entry Level Research Grade Inverted Microscope Data Sheet
Learn about the features of this simple, easy-to-use microscope that is ideal for education.
Fisherbrand Advanced Research Grade Upright Microscope Data Sheet
See the features that make this microscope ideal for the creation of publication-quality photos.
Fisherbrand Research Grade Upright Microscope Data Sheet
This microscope offers exceptional optical quality that makes it ideal for educational, clinical, and research labs.
Fisherbrand Entry Level Upright Microscope Data Sheet
Learn about the features that make this microscope perfect for both educational purposes and basic lab use.
More
Fisherbrand Research Grade Stereo Zoom Microscope Data Sheet
Learn about the features that make this microscope ideal for viewing macro-sized specimens and for use in college labs.
Fisherbrand Entry Level Stereo Zoom Microscope Data Sheet
See the features of this microscope, designed for the viewing of macro-sized specimens and ideal for use in college labs.
Fisherbrand Research Grade Stereo Microscope Data Sheet
Learn about the unique features that make this microscope ideal for high school classrooms.
Fisherbrand Entry Level Stereo Microscope Data Sheet
Find out more about this basic, fixed magnification stereo microscope, which includes features that make it perfect for simple applications.