Proteintech
Manufacturer Code:206971AP150UL
Catalog # 50-173-3041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hemimelic extra toes; HHG 1 HHG1 HLP3 HPE3 MCOPCB5 SHH SMMCI Sonic hedgehog protein TPT TPTPS; HHG-1; SHH; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; ShhN; ShhNC; ShhNp; short digits; sonic hedgehog homolog; Sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein N-product
Gene Aliases: 9530036O11Rik; Dsh; HHG1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; SHH; SMMCI; TPT; TPTPS; Vhh-1
UniProt ID: (Human) Q15465, (Mouse) Q62226, (Rat) Q63673
Entrez Gene ID: (Human) 6469, (Mouse) 20423, (Rat) 29499
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.