OriGene
Manufacturer Code:CF808337
Catalog # 50-166-8784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H8 |
Immunogen | Full length human recombit protein of human KRT16 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CK-16; CK16 FNEPPK K16 K1CP KRT16A NEPPK PC1; cytokeratin 16; Cytokeratin-16; focal non-epidermolytic palmoplantar keratoderma; K16; keratin 16, type I; Keratin, type I cytoskeletal 16; Keratin-16
Gene Aliases: CK16; FNEPPK; K16; K1CP; KRT16; KRT16A; NEPPK; PC1
UniProt ID: (Human) P08779
Entrez Gene ID: (Human) 3868
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.