Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone
Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-FMK is an irreversible and cell permeable inhibitor of CPP32/Apopain.
$463.00 - $1406.00
Specifications
Color | White |
---|---|
Content And Storage | Store in a cool, dry place. Dessicate. |
Product Type | Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone |
Molecular Weight (g/mol) | 668 |
Form | Solid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP3FK01005
|
MP Biomedicals™
03FK01005 |
5 mg |
Each for $1,406.00
|
|
|||||
MP3FK01001
|
MP Biomedicals™
03FK01001 |
1 mg |
Each for $463.00
|
|
|||||
MP3FK01003
|
MP Biomedicals™
03FK01003 |
3 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
A member of the CIE/CED-3 family of cysteine proteases.
Specifications
White | |
Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone | |
Solid |
Store in a cool, dry place. Dessicate. | |
668 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title