Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ White Biohazard Boot Cover
Click to view available options
Size:
Large
X-Large
Description
- Both fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and bloodborne pathogens
- Durable fabric is built to withstand tears and abrasions in tough working environments
- Taped seams add an extra layer of protection, helping prevent contaminants from entering or seeping through exposed seams
- Non-skid soles offer added traction to help prevent accidental slips
- Elastic ankle and top for a snug, secure fit
- Possible applications include emergency response, healthcare, category 3 black water, crime scene remediation, animal research, and more
Specifications
Size | Large |
Material | Non-woven Spunbond Polypropylene |
Latex Free | Yes |
Coating | None |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Top and Ankles |
Sole Type | Non-skid |
Height (English) | 17 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction