Learn More
Cytiva Whatman™ UniFilter™ 384-Well Microplates
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Supplier: Cytiva 77002110
Description
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
Can be used with gel filtration media for high-throughput sequencing reaction clean-up, including removal of dye blobs |
DNA Binding | |
Clear | |
384 | |
Long drip director |