Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ VU 0238429
Selective positive allosteric modulator of M5 receptors
$191.40 - $812.00
Specifications
Purity | 0.97 |
---|---|
Content And Storage | Store at -20°C |
Product Type | VU 0238429 |
Molecular Weight (g/mol) | 351.28 |
Inhibitors | VU 0238429 |
Description
Selective positive allosteric modulator of M5 receptors (EC50 values are 1.16, >30 and >30 μM at M5, M1 and M3 receptors respectively). Displays no activity at M2 and M4 receptors.Specifications
0.97 | |
VU 0238429 | |
VU 0238429 |
Store at -20°C | |
351.28 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title