Learn More
CTL Scientific Supply Corp VISOCOLOR ECO OXYGEN
SDP
Supplier:  CTL Scientific Supply Corp 931088
Encompass_Preferred
Environmental test kit for Oxygen Gradation of scale: 0 - 1 - 2 - 3 - 4 - 6 - 8 - 10 mg/l 50 tests/kit
Catalog No. 50-300-383