Learn More
Thomson Instrument Company Vacuum Manifold, Metal | Use w/ Filter Plates | EA
Supplier: Thomson Instrument Company 981802
Thomson Instrument Company high efficiency vacuum manifold can be connected directly to your house vacuum line and is designed for use with Thomson filter plates. These filter plates are offered in both 24 and 96-well configurations and have a long funnel design to eliminate cross-contamination between sample collection wells by inserting below the top of the collection plate. Thomson offers PVDF, PTFE and PP filter plates to be compatible with a wide variety of solvents and uses. The manifold will fit plates using SLAS specifications. Plates are individually wrapped for your convince. Thomson Instrument Company manufactures its products in the USA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.