Learn More
Foxx Life Sciences Vactrap™ Vacuum Trap System (Vacuum Flask Alternative) for Aspiration & Vacuum Protection, Shatterproof 2+1 Liter Aspirator Bottle Assembly with Secondary Container, Bleach Compatible HDPE, 2L Bottle with 1L Overflow Bottle & Inline Vent Filter
Supplier: Foxx Life Sciences 3022101FLS
Foxx Life Sciences' Vactrap is the world's most advanced vacuum trap system. Ideal for protection of vacuum lines and scientists alike, Vactrap safely contains waste from supernatant removal, chemical separation, and culture media aspiration. Best of all, Vactrap comes ready to use, simply attach to your vacuum and begin your work. The full Vactrap system includes two Foxx Vacuum Bottles (Autoclavable PP or Chemically-Resistant HDPE), each with a patented VersaCap closure for tangle-free opening, no need to disconnect tubing ever. A 0.2um PTFE inline vent filter retains any aerosolized bacteria, and the entire assembly is nested neatly in a secondary container with a small footprint and incredible stability. A pinch clamp allows for modulation of vacuum. Other Vactrap options offer one bottle, or setups without secondary containment. Say goodbye to glass. Say hello to Vactrap. Product Number: 302-2101-FLS Autoclavable: No, Autoclavable Alternative: 305-2101-FLS.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.