Learn More
Saint Gobain Performance Plastics VAC TUBING 1/4ID x 5/8 50FT/PK
Supplier: Saint Gobain Performance Plastics ACF00020
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Vacuum Sizes Diameter Inner 1/4 in. Diameter Outer 5/8 in. Thickness 3/16 in. Clear Maximum Pressure 53 psi at 73 deg.F