Learn More
MilliporeSigma™ Supelco™ Ultra-High Performance Fingertight Fittings, PEEK
The innovative line of Upchurch Ultra High Performance fittings are desigined to withstand extreme temperature anf pressures. This line of fittings is perfect for use within the increasingly demanding requirements for today's high performance analytical systems. The fittings are a 10/32 fingertight style for use with 1/16 in. and 1/32 in. OD tubing.
Supplier:  MilliporeSigma™ Supelco™ 51258U
 View more versions of this product
Catalog No. 11-101-0816
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Specifications
Specifications
10/Pk.
LiteTouch ferrule assembly 1/16 (black)
1/16 Tubing
PEEK
Product Suggestions
Product Suggestions