Learn More
Revvity Health Sciences Inc Ultra HiBlock Buffer 5X, 10 mL
Supplier: Revvity Health Sciences Inc TRF1011C
Formulation of 5X buffer: 125 mM HEPES, pH 7.4, 0.5% Casein, 5 mg/mL Dextran-500 K, 2.5% Triton X-100, 2.5% Blocking reagent, 2.5% BSA and 0.25% Proclin-300