Learn More
Saint Gobain Performance Plastics TUBING VINCON PVC 100FT/PK
Supplier: Saint Gobain Performance Plastics ABH02007
Saint-Gobain Vincon Flexible PVC Tubing Diameter Inner 1/8 in. 3.2 mm Diameter Outer 1/4 in. 6.4 mm Thickness 1/16 in. 1.6 mm Max. Operating Temperature 165 deg.F 74 deg.C Overall Length 100 ft. 30.5 m