Learn More
Malvern Panalytical TUBING F/WET DISPERSION,15M
Supplier:  Malvern Panalytical MAZ2013
Encompass
Standard Tubing for Wet Dispersion Units15 metres of standard tygon tubing (ST R-3603), for use with all Malvernlaser diffraction wet dispersion units.
Catalog No. NC2298569