Learn More
Saint Gobain Performance Plastics Tube Tygon 1/16 x 1/8 100FT/PK
Supplier: Saint Gobain Performance Plastics ACF02002
Saint-Gobain Tygon S3 E-3603 Flexible Tubing 100 ft. Diameter Inner 1/16 in. 1.6 mm Diameter Outer 1/8 in. 3.17 mm Thickness 1/32 in. 0.8 mm Overall Length 100 ft. 30.5 m Must order minimum quantity of 2.