Learn More
Chemglass Life Sciences 1mL Craig Tube, Glass Only
Supplier: Chemglass Life Sciences MW7401
1mL Craig Tube, Glass Only. Used for microscale recrystallizing of small quantities of reaction product. Consists of a glass outer tube with a non-uniform grind to purposely allow leakage.