Learn More
Ace Glass, Inc. Tube, pressure, 60mL, 150psi, 25.4mm OD, 30.5cm long, complete with#15 back-seal plug
Supplier: Ace Glass, Inc. 8648103
60mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 25.4mm O.D. x 30.5cm long.