Learn More
Ace Glass, Inc. Tube, pressure, 2mL, 150psi, 8mm OD, 17.8cm long, complete with#7 front-seal plug
Supplier: Ace Glass, Inc. 864821
2mL glass pressure tube with #7 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 8mm O.D. x 17.8cm long.