Learn More
Ace Glass, Inc. Tube, pressure, 1mL, 150psi, 8mm OD, 10.2cm long, complete with#7 front-seal plug
Supplier: Ace Glass, Inc. 864810
1mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 8mm O.D. x 10.2cm long.