Learn More
Ace Glass, Inc. Tube, pressure, 210mL, 150psi, 38.1mm OD, 30.5cm long, complete with #15 front-seal plunger valve
Supplier: Ace Glass, Inc. 864886
185mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 38.1mm O.D. x 30.5cm long.