Learn More
Ace Glass, Inc. Tube, pressure, 120mL, 150psi, 38.1mm OD, 17.8cm long, complete with #15 front-seal plunger valve with thermowell
Supplier: Ace Glass, Inc. 8648172
100mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve with Thermowell. Dimensions: 38.1mm O.D. x 17.8cm long.