Learn More
TraceCERT™ Elemental Impurities according to ICH Q3D Oral, Standard 1, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 19041100ML
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.Certified content incl. uncertainty and expiry date are given on the certificate.Specifications
7697-37-2 | |
Certified Reference Material | |
TraceCERT™ Analytical Standard |
100 mL | |
TraceCERT | |
Hg, 30 mg/L |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC