Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Timestrip™ PLUS Indicators
Record elapsed time at or above a specified temperature threshold in a simple and quick way
Supplier: MilliporeSigma™ Supelco™ 0816810EA
Description
- Small, self-adhesive labels that can be attached to or integrated into a product or device
- Manual activation button on top
- On window turns blue to confirm it is armed
- Blue color appears in the viewing window if a breach of the threshold temperature has occurred
- Cumulative duration of all breaches is shown by progress of blue dye along time markers
- Limited shelf life, use by expiration date on the label
- Time Accuracy: ±15% (under isothermal conditions)
- Recommended Storage: room temperature
- Produced under ISO 9001:2008
- Irreversible, single use
Specifications
8 Hr/8°C | |
8 Hour Run-Out Window (Calibrated (at 10°C)) | |
Recording Elapsed Time at or Above a Specified Temperature Threshold | |
8°C |
Aluminum/Plastic | |
40 x 19 mm | |
46°F | |
10/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Timestrip Plus is a trademark of Timestrip Limited
Spot an opportunity for improvement?
Provide Content Correction