Learn More
New England Biolabs, Inc. T7 RNA Polymerase (High Concentration) – 50000 units
Supplier: New England Biolabs, Inc. M0460T
T7 RNA Polymerase (High Concentration) is offered at a 20-fold higher concentration than our standard T7 RNA Polymerase and is ideal for experienced users interested in building and optimizing their own in vitro transcription reactions. The enzyme is accompanied by RNA Polymerase Reaction Buffer and magnesium chloride solution to further enable testing of different conditions such as nucleotide concentrations. For standard RNA synthesis and high yield reactions, we recommend our T7 RNA Polymerase and HiScribe kits. The HiScribe kits are specifically designed to produce high quality, high yield RNA in a short time with minimal optimization. In vitro transcription involves multiple components in addition to T7 RNA Polymerase including ribonucleotides, inorganic pyrophosphatase and RNase Inhibitor, which can be purchased separately. Bacteriophage T7 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the T7 phage promoters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.