Learn More
New England Biolabs, Inc. HiScribe™ T7 Quick High Yield RNA Synthesis Kit – 50 reactions
Supplier: New England Biolabs, Inc. E2050S
The HiScribe T7 Quick High Yield RNA Synthesis Kit is designed for quick set-up and production of large amounts of RNA in vitro. The reaction can be set up conveniently by combining the NTP buffer mix, T7 RNA Polymerase mix and a suitable DNA template. The kit also allows for capped RNA or dye-labeled RNA synthesis by incorporation of cap analog (ARCA, NEB #S1411) or dye-modified nucleotides. RNA synthesized with the kit can be used for RNA structure and function studies, ribozyme biochemistry, as probes for RNase protection assays and hybridization based blots, anti-sense RNA and RNAi experiments, microarray analysis and microinjection, as well as in vitro translation and RNA vaccines. To synthesize high specific activity radioactive RNA probes or RNA with 100% substitution of one or more modified nucleotides we recommend using the T7 High Yield RNA Synthesis Kit , in which the four nucleotides are supplied separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.