Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ System Reagents for Phadia™ 250 or Phadia™ 100
Replacement reagents provide true accuracy, high precision and outstanding quality
$38.97 - $107.03
Catalog Number | Mfr. No. | For Use With (Equipment) | Quantity | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Quantity | Type | Price | Quantity & Availability | |||
10-944-001P
|
Thermo Scientific™
10944001 |
Phadia 250 | ImmunoCAP: 6 x ∽315; EliA: 6 x >170 | Development Solution |
Pack of 6 for $100.00
|
|
|||
10-944-101P
|
Thermo Scientific™
10944101 |
Phadia 250 | ImmunoCAP: 6 x ∽200; EliA: 6 x >110 | Development Solution |
Pack of 6 for $107.03
|
|
|||
10-944-501P
|
Thermo Scientific™
10944501 |
Phadia 100 | 6 x 4 | ImmunoCAP Specific IgE Negative Control |
Pack of 24 for $38.97
|
|
|||
10-944-201P
|
Thermo Scientific™
10944201 |
Phadia 250 | ImmunoCAP: 6 x ∽185; EliA: 6 x >560 | Stop Solution |
Pack of 6 for $51.01
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-942-201P
|
Thermo Scientific™
10942201 |
Phadia 250 | ImmunoCAP: 6 x 1 l; EliA: 6 x 1 l | Washing Solution | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
10-947-901P
|
Thermo Scientific™
10947901 |
Phadia 250 | EliA: 6 x >292 | Stop Solution | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title