Learn More
World Precision Instrument Surgical Skin Stapler Handle & Cartridge
Supplier: World Precision Instrument WPSS01
The WPSS01 skin stapler is an affordable, reliable solution for veterinary wound closures. A skin stapler is designed for the efficient closure of surgical incisions or wounds by applying surgical staples to the skin, reducing surgery time and time under anesthesia. This handheld instrument features reloadable cassettes which are sold separately. The clear handle, which is autoclavable, allows you to spot contamination. The angled tip design gives you a clear view of the incision line. Pre-loaded, sterile staple cassettes containing 35 staples made loading efficient, and minimizes the chance of infection at the wound site. The lightweight and durable design is also ergonomic, making it easy to load and simple to use. The WPI skin stapler is made in the USA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.