Learn More
Sklar Instruments Forceps, Surgi-OR Jeweler Style Forceps, Sklar 4-1/2 in., Straight, Smooth, Size 5, Stainless Steel, EA.
Supplier: Sklar Instruments 95817
Forceps, Surgi-OR Jeweler Style Forceps, Sklar, 4-1/2 in., Straight, Smooth, Size 5, Stainless Steel, EA. Surgi-OR Jeweler Style Forceps are grasping forceps used in microsurgical procedures. They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/2 inches.Lifetime warranty against all manufacturer defects.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.