Learn More
Cole-Parmer Stuart SCF3/1/02 Tube Adaptors, 0.2 mL; 12/pk
Supplier: Cole-Parmer 8523004
The Stuart SCF3 Microcentrifuge includes a fixed-angle rotor that accommodates up to 12 tubes with a volume of 1.5- or 2.0-mL. Tube adapters to use 0.2- or 0.5-mL tubes can be ordered, as well as a strip tube rotor to accommodate two 8-place 0.2-mL strip tubes.