Learn More
Revvity Health Sciences Inc STK-Substrate 1-biotin, 50 g
Supplier:  Revvity Health Sciences Inc 61ST1BLE
Encompass
This substrate is a component of HTRF KinEASE S1 kit developed for an optimal kinase enzymatic step. It can be ordered as a spare part, e.g. when studying serine/threonine kinases with exceptionally high substrate Km.
Catalog No. 50-211-8287