Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stoelting™ Steam and EO Gas Indicator Strips
Steam and EO Gas Indicator Strips change color after sterilizing.
Catalog Number | Mfr. No. | Model | Length (English) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Length (English) | Includes | Price | Quantity & Availability | |||
10-000-705
|
Stoelting™
50991 |
Steam indicator strips | Short, 4 in. | Box of 250 |
Pack of 250 for $81.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-000-706
|
Stoelting™
50992 |
Long, 8 in. | EO gas indicator strips | Box of 240 |
Pack of 240 for $148.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-000-704
|
Stoelting™
50990 |
Steam indicator strips | Long, 8 in. | Box of 240 | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the Fisher Scientific channel. | N/A | |||
Description
- 8 inch strips are designed for use with large packs or may be torn in half for small packs
- 4 in. strips for smaller packs also available.
Specifications
Neuroscience |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title