Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SpeciMAX™ Dx Stabilized Saliva Collection Kit
Intended for the collection and preservation of saliva specimens in the presence of healthcare professionals as an aid in nucleic acid-based Polymerase Chain Reaction (PCR) for In Vitro Diagnostic purposes.
Supplier: Thermo Scientific™ A51023
Description
- Intended for the collection and preservation of saliva specimens in the presence of healthcare professionals
- An aid in nucleic acid-based polymerase chain reaction (PCR) for in vitro diagnostic purpose
- The individual single-use peel-pouch kit comes with an easy-to-use spill-resistant funnel, an automation-friendly saliva collection tube, and a screw cap to secure the sample after collection
- The SpeciMAX Dx saliva collection tube is pre-filled with 1 mL of stabilization agent for virus inactivation
- Integrates easily into automatic processing and analysis workflows that are part of high-throughput diagnostic testing
- Refrigeration or freezing stabilized saliva samples is not recommended as precipitation of the stabilization agent has downstream effects
Features of the SpeciMAX Dx Stabilized Saliva Collection Kit include:
- Inactivation of common respiratory viruses
- Stabilization solution that preserves gDNA, RNA, and viral nucleic acid in saliva samples
- Nonhazardous stabilization solution that is compatible with all disinfection protocols—no reaction with bleach (no guanidinium or ethanol)
- Barcoded, standardized tubes and caps that fit automated workflows
- Instrumental in high-throughput diagnostic testing
Sample storage
- Refrigeration or freezing the stabilized saliva samples is not recommended as precipitation of the stabilization agent has downstream effects
- The SpeciMAX stabilization agent preserves nucleic acid in saliva samples at ambient temperature for the following lengths of time:
- gDNA: 6 months
- RNA: 30 days
- Viral nucleic acid: 30 days
Easy to use
- Each SpeciMAX Dx Stabilized Saliva Collection Kit comes ready to use in individually packaged easy-peel blister pouches
- To use the kit, the saliva donor follows four easy steps:
- Remove and save the cap, then attach the funnel securely to the collection tube.
- Fill saliva to the 2.0 line (not including bubbles)
- Unscrew and discard the funnel (do not reuse)
- Secure the cap on the tube, then mix by inverting the tube five times
- For best results, saliva donors should be instructed to refrain from eating, drinking, using tobacco products, or chewing gum for at least 30 minutes prior to sample collection
Specifications
Stabilized Saliva Collection Kit | |
• Virus inactivating and nucleic stabilization solution (1 mL) • Pre-filled saliva collection tube, cap, and funnel Store at 15&°C to 30°C. |
|
Virus inactivating and nucleic stabilization solution (1 mL) pre-filled saliva collection tube, cap, and funnel | |
Non-sterile | |
6 mL |
CE, IVD | |
For In Vitro Diagnostic Use | |
100 Kits | |
Store at 15°C to 30°C |
Safety and Handling
ShelfLife : 2 years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction