Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel DMAP, 40 to 63 μm, 60 Å
Our DMAP is a heterogeneous reagent and organic scavenger for trapping acyl chlorides and sulfonyl chlorides.
$247.00 - $11400.00
Specifications
Physical Form | Irregular |
---|---|
For Use With (Application) | Post-reaction Mixture or in Flash Chromatography |
Packaging | HDPE Bottle |
Particle Size | 40 to 63 μm |
Specific Surface Area | 500 m2/g |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-184-410
|
SiliCycle™
R75630B10G |
10 g |
Each for $247.00
|
|
|||||
01-184-411
|
SiliCycle™
R75630B25G |
25 g |
Each for $475.50
|
|
|||||
01-184-412
|
SiliCycle™
R75630B50G |
50 g |
Each for $891.50
|
|
|||||
01-184-413
|
SiliCycle™
R75630B100G |
100 g |
Each for $1,425.00
|
|
|||||
01-184-435
|
SiliCycle™
R75630B250G |
250 g |
Each for $3,360.00
|
|
|||||
01-184-414
|
SiliCycle™
R75630B500G |
500 g |
Each for $6,350.00
|
|
|||||
01-184-415
|
SiliCycle™
R75630B1KG |
1 kg |
Each for $11,400.00
|
|
|||||
01-184-409
|
SiliCycle™
R75630B5G |
5 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
SiliaBond™ DMAP acts an Organic Scavenger for Electrophiles (Covalent Bonding for Acyl Chlorides and Sulfonyl Chlorides). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity.
- Great variety of formats for all purifications scales, from laboratory to process to plant scales
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Irregular | |
HDPE Bottle | |
500 m2/g | |
Bulk Functionalized Irregular Silica Gel | |
Bulk or Glass Gravity Columns | |
DMAP | |
SiliaBond™ |
Post-reaction Mixture or in Flash Chromatography | |
40 to 63 μm | |
0.674 g/mL | |
Yes | |
Irregular | |
60 Å | |
DMAP |
Safety and Handling
Recommended Storage : Keep Cool (<8°C), Dry and Under Inert Atmosphere
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.