Learn More
Sestrin 2 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc CL59410795100UL
This item is not returnable.
View return policy
Description
The Sestrin 2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Sestrin 2. This antibody recognizes human, mouse, rat antigen. The Sestrin 2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Sestrin 2 | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Mouse, Rat |
Immunofluorescence | |
CoraLite 594 | |
100 μL | |
83667 | |